Assay Details
Gene Symbol: LOC100284118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157015.1 | NP_001150487.1 | 438 | 101 | ||
GenBank mRNA | BT038766.1 | - | 438 | 101 | ||
BT054479.1 | - | 347 | 101 | |||
BT060497.1 | - | 401 | 101 | |||
BT064091.1 | - | 449 | 101 | |||
EU967137.1 | - | 547 | 101 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
