Assay Details
Gene Symbol: LOC100284118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157015.1 | NP_001150487.1 | 639 | 68 | ||
GenBank mRNA | BT038766.1 | - | 639 | 68 | ||
BT054479.1 | - | 548 | 68 | |||
BT060497.1 | - | 630 | 68 | |||
BT064091.1 | - | 650 | 68 | |||
EU967137.1 | - | 748 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
