Assay Details
Gene Symbol: BE997321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157913.1 | NP_001151385.1 | 735 | 101 | ||
GenBank mRNA | EU970525.1 | - | 735 | 101 |
Gene Symbol: LOC100381822 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001174617.1 | NP_001168088.1 | 270 | 101 | ||
GenBank mRNA | BT062356.1 | - | 270 | 101 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
