Assay Details
Gene Symbol: LOC100285873 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158763.1 | NP_001152235.1 | 339 | 86 | ||
GenBank mRNA | BT065615.1 | - | 278 | 86 | ||
EU974427.1 | - | 339 | 86 | |||
EU975080.1 | - | 330 | 86 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
