Assay Details
| Gene Symbol: LOC100285873 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158763.1 | NP_001152235.1 | 402 | 91 | ||
| GenBank mRNA | BT065615.1 | - | 341 | 91 | ||
| EU974427.1 | - | 402 | 91 | |||
| EU975080.1 | - | 393 | 91 | |||
More Information
Set Membership: |
|

LOC100285873
Genomic Map