Assay Details
| Gene Symbol: cha101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111617.1 | NP_001105087.1 | 1611 | 101 | ||
| GenBank mRNA | AF461813.1 | - | 1611 | 101 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cha101| Gene Symbol: cha101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111617.1 | NP_001105087.1 | 1611 | 101 | ||
| GenBank mRNA | AF461813.1 | - | 1611 | 101 | ||
Set Membership: |
|