Assay Details
| Gene Symbol: pox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111674.1 | NP_001105144.1 | 739 | 80 | ||
| GenBank mRNA | AJ401274.1 | - | 739 | 80 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pox1| Gene Symbol: pox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111674.1 | NP_001105144.1 | 739 | 80 | ||
| GenBank mRNA | AJ401274.1 | - | 739 | 80 | ||
Set Membership: |
|