Assay Details
| Gene Symbol: hag101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111675.1 | NP_001105145.1 | 599 | 67 | ||
| GenBank mRNA | AF440227.1 | - | 614 | 67 | ||
| AJ428540.1 | - | 628 | 67 | |||
| BT063893.1 | - | 576 | 67 | |||

hag101
Genomic Map