Assay Details
| Gene Symbol: gdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111831.1 | NP_001105301.1 | 640 | 66 | ||
| GenBank mRNA | AY106054.1 | - | 655 | 66 | ||
| D49475.1 | - | 640 | 66 | |||
| U93560.1 | - | 520 | 66 | |||
| U93561.1 | - | 520 | 66 | |||
More Information
Set Membership: |
|

gdh1
Genomic Map