Assay Details
| Gene Symbol: IDP2557 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136697.1 | NP_001130169.1 | 750 | 84 | ||
| GenBank mRNA | BT033305.1 | - | 750 | 84 | ||
| BT042131.1 | - | 774 | 84 | |||
More Information
Set Membership: |
|

IDP2557
Genomic Map