Assay Details
| Gene Symbol: LOC100191518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136950.1 | NP_001130422.1 | 1332 | 80 | ||
| GenBank mRNA | BT033618.1 | - | 1332 | 80 | ||
| BT066940.1 | - | 1333 | 80 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100191518| Gene Symbol: LOC100191518 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136950.1 | NP_001130422.1 | 1332 | 80 | ||
| GenBank mRNA | BT033618.1 | - | 1332 | 80 | ||
| BT066940.1 | - | 1333 | 80 | |||