Assay Details
| Gene Symbol: LOC100191631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137060.1 | NP_001130532.1 | 1945 | 97 | ||
| GenBank mRNA | BT033779.1 | - | 1945 | 97 | ||
| BT067439.1 | - | 1931 | 97 | |||
More Information
Set Membership: |
|

LOC100191631
Genomic Map