Assay Details
Gene Symbol: LOC100191631 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137060.1 | NP_001130532.1 | 1616 | 59 | ||
GenBank mRNA | BT033779.1 | - | 1616 | 59 | ||
BT067439.1 | - | 1602 | 59 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
