Assay Details
Gene Symbol: pco071744(596) | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137073.1 | NP_001130545.1 | 1007 | 92 | ||
GenBank mRNA | BT033799.1 | - | 1007 | 92 | ||
BT067543.1 | - | 913 | 92 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
