Assay Details
| Gene Symbol: pco071744(596) | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137073.1 | NP_001130545.1 | 1181 | 93 | ||
| GenBank mRNA | BT033799.1 | - | 1181 | 93 | ||
| BT067543.1 | - | 1087 | 93 | |||
More Information
Set Membership: |
|

pco071744(596)
Genomic Map