Assay Details
Gene Symbol: spr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137102.1 | NP_001130574.1 | 1396 | 92 | ||
GenBank mRNA | BT033842.1 | - | 1396 | 92 | ||
BT042981.1 | - | 1411 | 92 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
