Assay Details
Gene Symbol: cad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137309.1 | NP_001130781.1 | 844 | 70 | ||
GenBank mRNA | BT034128.1 | - | 844 | 70 | ||
BT035050.1 | - | 766 | 70 | |||
BT040774.1 | - | 841 | 70 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
