Assay Details
Gene Symbol: sad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137318.1 | NP_001130790.1 | 1606 | 69 | ||
GenBank mRNA | BT034140.1 | - | 1606 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
