Assay Details
| Gene Symbol: LOC100191972 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137396.1 | NP_001130868.1 | 831 | 79 | ||
| GenBank mRNA | BT034234.1 | - | 831 | 79 | ||
| BT054525.1 | - | 850 | 79 | |||
| BT061240.1 | - | 909 | 79 | |||
| EU676041.1 | - | 599 | 79 | |||
More Information
Set Membership: |
|

LOC100191972
Genomic Map