Assay Details
Gene Symbol: LOC100192094 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137517.1 | NP_001130989.1 | 630 | 79 | ||
GenBank mRNA | BT034400.1 | - | 544 | 79 | ||
BT063300.1 | - | 528 | 79 | |||
EU964300.1 | - | 630 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
