Assay Details
Gene Symbol: TIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146952.1 | NP_001140424.1 | 978 | 69 | ||
GenBank mRNA | BT038799.1 | - | 978 | 69 | ||
BT066543.1 | - | 791 | 69 | |||
EU952625.1 | - | 806 | 69 | |||
EU976695.1 | - | 902 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
