Assay Details
Gene Symbol: LOC100274370 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001148730.1 | NP_001142202.1 | 492 | 69 | ||
GenBank mRNA | BT042871.1 | - | 492 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
