Assay Details
Gene Symbol: LOC100275133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001149302.1 | NP_001142774.1 | 203 | 76 | ||
GenBank mRNA | EU953579.1 | - | 203 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
