Assay Details
Gene Symbol: LOC100275133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001149302.1 | NP_001142774.1 | 643 | 111 | ||
GenBank mRNA | EU953579.1 | - | 643 | 111 | ||
EU954440.1 | - | 744 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |
