Assay Details
| Gene Symbol: LOC100280498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153418.1 | NP_001146890.1 | 895 | 89 | ||
| GenBank mRNA | EU952098.1 | - | 895 | 89 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100280498| Gene Symbol: LOC100280498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153418.1 | NP_001146890.1 | 895 | 89 | ||
| GenBank mRNA | EU952098.1 | - | 895 | 89 | ||