Assay Details
Gene Symbol: LOC100281512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154430.1 | NP_001147902.1 | 898 | 59 | ||
GenBank mRNA | EU956950.1 | - | 898 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
