Assay Details
Gene Symbol: LOC100283984 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156882.1 | NP_001150354.1 | 1104 | 60 | ||
GenBank mRNA | EU966657.1 | - | 1104 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
