Assay Details
Gene Symbol: pks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157069.1 | NP_001150541.1 | 857 | 78 | ||
GenBank mRNA | EU967359.1 | - | 857 | 78 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
