Assay Details
Gene Symbol: pks1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157069.1 | NP_001150541.1 | 1007 | 103 | ||
GenBank mRNA | EU967359.1 | - | 1007 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
