Assay Details
Gene Symbol: LOC100284209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157104.1 | NP_001150576.1 | 211 | 63 | ||
GenBank mRNA | EU967508.1 | - | 211 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
