Assay Details
Gene Symbol: LOC100284240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157135.1 | NP_001150607.1 | 445 | 87 | ||
GenBank mRNA | EU967616.1 | - | 445 | 87 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
