Assay Details
Gene Symbol: LOC100284726 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157621.1 | NP_001151093.1 | 853 | 92 | ||
GenBank mRNA | EU969472.1 | - | 853 | 92 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
