Assay Details
| Gene Symbol: mde1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158276.1 | NP_001151748.1 | 462 | 76 | ||
| GenBank mRNA | EU972095.1 | - | 462 | 76 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

mde1| Gene Symbol: mde1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158276.1 | NP_001151748.1 | 462 | 76 | ||
| GenBank mRNA | EU972095.1 | - | 462 | 76 | ||