Assay Details
Gene Symbol: LOC100272398 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146878.1 | NP_001140350.1 | 224 | 89 | ||
GenBank mRNA | BT038636.1 | - | 224 | 89 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
