Assay Details
Gene Symbol: LOC100281236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154155.1 | NP_001147627.1 | 552 | 101 | ||
GenBank mRNA | EU956037.1 | - | 552 | 101 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
