Assay Details
| Gene Symbol: LOC100281373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154291.1 | NP_001147763.1 | 330 | 71 | ||
| GenBank mRNA | EU956496.1 | - | 330 | 71 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100281373| Gene Symbol: LOC100281373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154291.1 | NP_001147763.1 | 330 | 71 | ||
| GenBank mRNA | EU956496.1 | - | 330 | 71 | ||