Assay Details
Gene Symbol: LOC100282198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001155110.1 | NP_001148582.1 | 641 | 101 | ||
GenBank mRNA | EU960004.1 | - | 641 | 101 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
