Assay Details
Gene Symbol: LOC100282198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001155110.1 | NP_001148582.1 | 786 | 89 | ||
GenBank mRNA | EU960004.1 | - | 786 | 89 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
