Assay Details
Gene Symbol: LOC100283755 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156654.1 | NP_001150126.1 | 710 | 56 | ||
GenBank mRNA | EU965835.1 | - | 710 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
