Assay Details
Gene Symbol: LOC100285223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158117.1 | NP_001151589.1 | 921 | 101 | ||
GenBank mRNA | EU971336.1 | - | 921 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
