Assay Details
Gene Symbol: LOC100285500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158392.1 | NP_001151864.1 | 1055 | 68 | ||
GenBank mRNA | BT054828.1 | - | 1042 | 68 | ||
EU972543.1 | - | 1055 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
