Assay Details
Gene Symbol: LOC100286167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159055.1 | NP_001152527.1 | 443 | 87 | ||
GenBank mRNA | BT087323.1 | - | 431 | 87 | ||
EU975924.1 | - | 443 | 87 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
