Assay Details
Gene Symbol: LOC100272252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146747.1 | NP_001140219.1 | 1030 | 100 | ||
GenBank mRNA | BT038351.1 | - | 1030 | 100 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
