Assay Details
Gene Symbol: LOC100281196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154115.1 | NP_001147587.1 | 2151 | 67 | ||
GenBank mRNA | EU955884.1 | - | 2151 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
