Assay Details
Gene Symbol: adk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154161.1 | NP_001147633.1 | 595 | 77 | ||
GenBank mRNA | EU955541.1 | - | 622 | 77 | ||
EU956082.1 | - | 595 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
