Assay Details
Gene Symbol: LOC100281252 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154171.1 | NP_001147643.1 | 751 | 90 | ||
GenBank mRNA | EU956116.1 | - | 751 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
