Assay Details
Gene Symbol: LOC100281405 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154323.1 | NP_001147795.1 | 2174 | 80 | ||
GenBank mRNA | BT061656.1 | - | 2174 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
