Assay Details
Gene Symbol: LOC100282579 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001155487.1 | NP_001148959.1 | 647 | 65 | ||
GenBank mRNA | EU961527.1 | - | 647 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
