Assay Details
Gene Symbol: LOC100283039 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001155941.1 | NP_001149413.1 | 794 | 88 | ||
GenBank mRNA | BT067623.1 | - | 236 | 88 | ||
BT069508.1 | - | 794 | 88 | |||
EU963245.1 | - | 248 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
