Assay Details
Gene Symbol: LOC100283039 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001155941.1 | NP_001149413.1 | 1400 | 65 | ||
GenBank mRNA | BT067623.1 | - | 842 | 65 | ||
BT069508.1 | - | 1400 | 65 | |||
EU963245.1 | - | 854 | 65 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
